Copper(I) sulfide, 99.5% (metals basis), Alfa Aesar

For Methods: Luminous Paints, Electrodes, Solar Cells

Overview
Brand: Alfa Aesar

Manufacturer Part Number: 014718.A1

COPPER(I) SULFIDE, 99.5% 1KG

UNSPSC: 12141711

Code: NEW

Additional Details:
Additional Details: CAS Number: 22205-45-4 Weight: 0.01000kg


Quantity 1kg

Product Code. 11306489

Quantity Price
1 £ 515.69 / 1kg
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Copper(I) sulfide
CAS 22205-45-4
Physical Form Powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Cu2S
Formula Weight 159.14
MDL Number MFCD00016062

Specification

Formula Weight 159.14
Quantity 1kg
Color Black
Chemical Name or Material Copper(I) sulfide
EINECS Number 244-842-9
Odor Odorless
Physical Form Powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Cu2S
Appearance Black powder
MDL Number MFCD00016062
CAS 22205-45-4