missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper(I) selenide, 99.5% (metals basis)

Brand:  Alfa Aesar™ 045660.09

Code : NEW

Additional Details : CAS Number : 20405-64-5 Weight : 0.01000kg Transport : UN number : 3077 Chem class : 9 Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 15472427

  • £69.30 / 10g


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Chemical Identifiers

Copper (I) selenide
Cu2Se
MFCD00049908
14,2666
20405-64-5
206.05
UN3077

Specifications

Copper (I) selenide
Pieces
(metals basis)
206.05
UN3077
6.84g/mL
Dark blue to black
10g
20405-64-5
99.5%
Cu2Se
MFCD00049908
14,2666
2mm & down
1113°C
Videos
SDS