missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) phenylacetylide, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
2 g
Description
Copper(I) phenylacetylide participates in efficient coupling with a range of aryl iodides and bromides to give substituted tolanes which, if possessing an ortho amino or hydroxy group, can be thermolyzed to yield 2-phenylbenzofurans, -indoles, and -furo[3,2-b]pyridines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Quantity | 500 mg |
UN Number | UN3088 |
Sensitivity | Air & Moisture Sensitive |
Solubility Information | Soluble in acetone and acetonitrile; Insoluble in water |
Formula Weight | 153.68 |
Physical Form | Powder |
Chemical Name or Material | Copper(I) phenylacetylide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'