missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) oxide, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Description
Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Purple-Red |
Physical Form | Powder |
Quantity | 2.5 kg |
Assay Percent Range | 99% (Metal basis) |
Formula Weight | 143.08 |
Odor | Odorless |
Appearance | Purple-red powder |
Chemical Name or Material | Copper(I) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'