missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) iodide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407429
Click to view available options
Quantity:
5 g
25 g
100 g
7681-65-4
CuI
MFCD00010978
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
7681-65-4
CuI
MFCD00010978
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

Light, Air and Moisture Sensitive

Copper(I) iodide is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point ∼1290°C
Melting Point 605°C
Color Brown to White
Physical Form Chunks and/or Powder
Quantity 5 g
Assay Percent Range 99.998% (Metal basis)
Solubility Information Copper(I) iodide is soluble in ammonia,thiosulfate,sodium and potassium iodide solutions. Insoluble in water and dilute acids.
Formula Weight 190.45
Odor Weak odor
Refractive Index 2.346
Sensitivity Air, light, and moisture sensitive
Appearance White to pale brown chunks/powder
Density 5.63 g/cm3
Chemical Name or Material Copper(I) Iodide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'