missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) iodide, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Catalog Number p-2691235
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
7681-65-4
CuI
190.45
MFCD00010978
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g
7681-65-4
CuI
190.45
MFCD00010978
cuprous iodide, copper l iodide, copper i iodide metals basis, marshite, copper i iodide 250g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, for ultra trace analysis
TRUSTED_SUSTAINABILITY
Boiling Point 1290°C
Melting Point 605°C
Color Brown or Gray to White
Physical Form Powder
Quantity 5 g
Assay Percent Range 31.0 to 34% (Cu) (Complexometry)
Linear Formula CuI
Fieser 01,169; 02,92; 03,69; 05,167; 06,147; 07,81; 08,121; 09,124; 10,107; 11,141; 12,141; 16,98; 17,88
Merck Index 15, 2652
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ammonia and potassium iodide solutions,practically insoluble in alcohol
Formula Weight 190.45
Percent Purity 100.00%
Grade Trace Metal Basis
Packaging Glass bottle
Density 5.62
Chemical Name or Material Copper(I) iodide, 99.995%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(I) iodide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'