missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper(I) hexafluoro-2,4-pentanedionate 1,5-cyclooctadiene complex

Used in MOCVD as a source of Cu2O

Brand:  Alfa Aesar™ 041916.04

Code : NEW

Additional Details : CAS Number : 86233-74-1 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11394887

  • £130.80 / 2g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Copper(I) hexafluoro-2,4-pentanedionate 1,5-cyclooctadiene complex
378.77
Copper(I) hexafluoroacetylacetonate complex with 1,5-cyclooctadiene
86233-74-1
MFCD00156517

Specifications

Copper(I) hexafluoro-2,4-pentanedionate 1,5-cyclooctadiene complex
Powder/Aggregate
MFCD00156517
Soluble in THF, hexane
Yellow-green
2g
86233-74-1
378.77
Copper(I) hexafluoroacetylacetonate complex with 1,5-cyclooctadiene
60°C (0.1mmHg) (subl.)
93° to 100°C
Videos
SDS