missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) cyanide, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
2 x 2.5 kg
Description
Copper(I) cyanide is commonly used as a polymerization catalyst. The compound is useful in organic synthesis, as a catalyst, reagent in the preparation of nitriles(sandmeyer reaction). It is also used in electroplating copper and iron, silver plating, brass plating, and copper-tin alloy plating. It is used to make phthalocyanine dyes and pigments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 473°C |
Color | White |
Physical Form | Powder |
Quantity | 2 x 2.5 kg |
Beilstein | 3587244 |
Solubility Information | Practically insoluble in water and alcohol. Soluble in ammonium hydroxide,aqueous ammonia,pyridine and N-methylpyrrolidone. |
Formula Weight | 89.56 |
Odor | Characteristic |
Density | 2.92 |
Chemical Name or Material | Copper(I) cyanide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'