missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) cyanide, 99%, extra pure, ACROS Organics™

£32.35 - £270.00

Chemical Identifiers

CAS 544-92-3
Molecular Formula CCuN
Molecular Weight (g/mol) 89.56
MDL Number MFCD00010975
InChI Key DOBRDRYODQBAMW-UHFFFAOYSA-N
Synonym copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
PubChem CID 11009
IUPAC Name copper(1+);cyanide
SMILES [C-]#N.[Cu+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10012790
SDS View Documents
Acros Organics
202082500
250g Plastic bottle
£32.35
250g
View alternative products
Save Up to 
»
Estimated Shipment
26-01-2021
Please to purchase this item. Need a web account? Register with us today!
10583371
SDS View Documents
Acros Organics
202080010
1kg Plastic bottle
£84.59
1kg
View alternative products
Save Up to 
»
Estimated Shipment
01-02-2021
Please to purchase this item. Need a web account? Register with us today!
10454985
SDS View Documents
Acros Organics
202080051
5kg Plastic bottle
£270.00
5kg
View alternative products
Save Up to 
»
Estimated Shipment
28-01-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C-X bond formation reagent
Specifications

Chemical Identifiers

544-92-3
89.56
DOBRDRYODQBAMW-UHFFFAOYSA-N
11009
[C-]#N.[Cu+]
CCuN
MFCD00010975
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
copper(1+);cyanide

Specifications

Copper(I) cyanide
98.5% min. (ex Cu)
CuCN
05,166; 06,146; 09,124; 16,97
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies,practically insoluble in alcohol
[C-]#N.[Cu+]
89.56
89.56
99%
2.9200g/mL
20ppm max.
Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies, practically insoluble in alcohol
10ppm max.
474.0°C
544-92-3
CCuN
MFCD00010975
15, 2651
10ppm max.
DOBRDRYODQBAMW-UHFFFAOYSA-N
copper(1+);cyanide
11009
Powder
Extra Pure
100ppm max.
Plastic bottle
2.92
Beige-White to Green
250g
Safety and Handling

Safety and Handling

Product Identifier
  • Copper(I) cyanide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

SDS