missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 97%, pure, Thermo Scientific Chemicals

Catalog Number 10668124
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
7758-89-6
ClCu
99
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
7758-89-6
ClCu
99
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1490.0°C
Melting Point 430.0°C
Color Gray-Green
Physical Form Powder
Quantity 2.5 kg
Assay Percent Range 96% min. (Complexometry)
Linear Formula CuCl
Fieser 01,166; 02,91; 03,67; 04,109; 05,164; 06,145; 07,80; 08,118; 09,123; 11,140; 12,141; 13,152; 14,100; 15,101; 16,97
Merck Index 15, 2650
Solubility Information Solubility in water: 0.06g/L (25°C). Other solubilities: soluble in conc. hcl and nh4oh,forming complexes,practically insoluble in alcohol and acetone
Formula Weight 99
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Specific Gravity 4.14
Density 4.1400g/mL
Chemical Name or Material Copper(I) chloride, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(I) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'