missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Chloride, ≥97.0%, Honeywell™ Fluka™

Product Code. p-7120340 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15600880

Brand: Honeywell-Fluka 08253250G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7758-89-6
Molecular Formula ClCu
Molecular Weight (g/mol) 98.996
MDL Number MFCD00010971
InChI Key OXBLHERUFWYNTN-UHFFFAOYSA-M
Synonym cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
PubChem CID 62652
ChEBI CHEBI:53472
IUPAC Name chlorocopper
SMILES Cl[Cu]

Specifications

Boiling Point 1490°C
Melting Point 430°C
Color Gray
pH 5.0 (at 50.00g/L 20°C)
Quantity 250 g
Linear Formula CuCl
UN Number UN2802
Solubility Information Slightly soluble in water, 0.47g/L at 20°C (USP brand)
Formula Weight 99g/mol
Percent Purity ≥97.0%
Packaging Poly Bottle
Appearance Small beads
Vapor Pressure 1.3mmHg (546°C)
Density 3.530g/cu.cm
Chemical Name or Material Copper(I) Chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.