missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
The most notable physical characteristics of germania (GeO2) are its high index of refraction and its low optical dispersion. These make it especially useful for wide-angle camera lenses, microscopy, and for the core part of optical fibers. Germanium-on-insulator substrates are seen as a potential replacement for silicon on miniaturized chips.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Pieces |
Quantity | 50 g |
UN Number | UN3283 |
Solubility Information | Insoluble in water. |
Formula Weight | 215.12 |
Percent Purity | 99.99% |
Assay | (metals basis) |
Chemical Name or Material | Copper germanium selenide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'