missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Copper germanide, 99.99% (metals basis)

Brand:  Alfa Aesar™ 045604.09

Code : NEW

Additional Details : Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 15442407

  • £165.20 / 10g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Copper germanide
CuGe
136.16
MFCD12546178

Specifications

Copper germanide
99.99%
CuGe
MFCD12546178
10g
Pieces
(metals basis)
136.16
3 to 6mm
Videos
SDS