missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper foil, 1.0mm (0.04in) thick, 99.99% (metals basis), Thermo Scientific Chemicals
31.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 x 50 mm
50 x 250 mm
50 x 1000 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Copper colored foil |
Assay Percent Range | 99.9999% (Metals basis) |
Quantity | 50 x 50 mm |
Odor | Odorless |
Vapor Pressure | 0 hPa |
Form | Foil |
Chemical Name or Material | Copper |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'