missing translation for 'onlineSavingsMsg'
Learn More

Copper foil, 1.27mm (0.05in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7049218
Click to view available options
Quantity:
20 x 20 cm
20 x 100 cm
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 20 x 20 cm
Percent Purity 99.9%
Odor Odorless
Weight ≈455g/20x20cm
Form Foil
Assay (metals basis)
Chemical Name or Material Copper foil, 1.27mm (0.05 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'