Learn More
Alfa Aesar™ Copper foil, 0.025mm (0.001 in.) thick, annealed, uncoated, 99.8% (metals basis)
For Graphene growth
Brand: Alfa Aesar™ 046365.RG
Code : NEW
Additional Details : CAS Number : 7440-50-8 Weight : 0.01000kg
Quantity | 30x150cm |
---|
Chemical Identifiers
Copper foil | |
7440-50-8 | |
0.025mm (0.001 in.) thick, annealed, uncoated | |
MFCD00010965 |
Specifications
Copper foil | |
30x150cm | |
7440-50-8 | |
99.8% | |
MFCD00010965 | |
Copper |
≈20.2g/30x30cm | |
0.025mm (0.001 in.) thick, annealed, uncoated | |
Foil | |
(metals basis) | |
Odorless |