missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Copper Beryllium rod, alloy C17200, 9.525mm (0.375 in.) dia.

Brand:  Thermo Scientific Alfa Aesar 042483.BU

Copper Beryllium rod, alloy C17200, 9.525mm (0.375 in.) dia.

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15456485

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.