Learn More
Copper Beryllium foil, alloy C172, 0.13mm (0.005in) thick, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 041911.GU
86.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Uses include springs of all kinds where high strength and corrosion resistance are important, resistance welding electrodes, dies and high strength, corrosion resistant tubing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Copper Beryllium foil, Alloy C172 | |
Cu:Be; 98.1:1.9 wt% | |
150 x 200 mm | |
Odorless | |
Foil |
0.13mm (0.005 in.) thick, Temper: half hard | |
MFCD01320550 | |
Insoluble in water. | |
≈32.6g/150x200mm |
RUO – Research Use Only