missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper based methanol synthesis catalyst, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Description
Lurgi-low pressure process for methanol synthesis also uses a copper based catalyst which is CuO/ZnO.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown to Black |
Quantity | 1 kg |
UN Number | UN3077 |
Physical Form | Pellets |
Chemical Name or Material | Copper based methanol synthesis catalyst |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'