missing translation for 'onlineSavingsMsg'
Learn More

Copper, 99.9998%, (trace metal basis), Thermo Scientific™

Catalog Number p-4228256
Click to view available options
Quantity:
25 g
125 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For ultra trace analysis
TRUSTED_SUSTAINABILITY
Color Brown-Rust
Melting Point 1083.0°C
Boiling Point 2595.0°C
Assay Percent Range 99.9998%
Linear Formula Cu
Quantity 25 g
Solubility Information Solubility in water: insoluble.
Formula Weight 63.54
Percent Purity 99.9998%
Grade Trace Metal Basis
Packaging Glass bottle
Form Shot
Chemical Name or Material Copper
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'