missing translation for 'onlineSavingsMsg'
Learn More

Nylon Flocked Dry Swabs in Peel Pouches

Permits complete, rapid elution by keeping the entire sample close to the surface. Copan Diagnostics Nylon Flocked Dry Swabs in Peel Pouches are anatomically designed swabs to optimize analyte collection efficiency with improved patient comfort.

Brand:  Thermo Scientific Sterilin 503CS01

Code : 04

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 12769915

  • £136.00 / Pack of 100


In Stock 17
Add to basket

Description

Description

Sterile, individually wrapped swabs in a plastic applicator.
  • Ergonomic and Anatomic Design: Improves patient comfort and specimen collection
  • Rapid Automatic Elution: Instant and spontaneous release of sample into liquid media
  • Measurable and Consistant Uptake and Transfer: Quantitative measurement and improved test sensitivity
  • Patient Friendly: Anatomical design and a soft texture for rapidly and efficiently dislodging cells
  • Compatible with Multiple Applications: Rapid antigen testing, EIA, molecular-based assays, DFA, cytology testing, bacteriology and virology culture
  • Safe and Convenient: Molded breakpoint
Specifications

Specifications

Individually-wrapped in peel pouch
Yes
100mm
Plastic
Flexible minitip flocked
Specimen Collection Swab
Videos