missing translation for 'onlineSavingsMsg'
Learn More

Corning™ CoolRack™ M-PF Conical Tube Modules

Ensures ±0.1°C temperature uniformity across all tubes when cooling, snap freezing, heating or thawing.

Brand:  Corning™ 432048

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15532821

  • £301.74 / Each

Estimated Shipment
Add to basket



  • Modules eliminate variability originating from tubes placed directly into ice, dry ice, alcohol baths, water baths and other temperature sources
  • Place module directly on temperature source between −196° to >100°C and it will rapidly adapt to that temperature
  • All modules may be autoclaved, high-heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents
  • Applications include cooling reagents such as restriction enzymes, dNTPs and antibodies, alcohol-free dry ice snap freezing of tissue, virus and bacteria samples and bench top cryogenic tube sorting in liquid nitrogen


Conical Tube Thermoconductive Module
30 x 1.5mL conical tubes