Learn More
Miele Professional™ Conversion kit UBS 2
Conversion kit UBS 2 for use with selected series G 78 upper baskets in PG 85 machines
Brand: Miele Professional™ 10131100
Code : NEW
Additional Details : Weight : 0.73000kg
Description
- Converts old injector baskets to fit into new Miele PG85 range lab washing machines
- For load carriers and upper baskets for G 78 model series without drying connection
Specifications
Black |