missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Convection and Universal Oven Subframe

Use these frames with Memmert convection ovens. Memmert™ Convection and Universal Oven Subframes are adjustable in height to fit your needs.

£231.69 - £473.92

Specifications

Type Subframe
View More Specs

Products 6
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
13488846 Memmert™
B29750
Models 110 and 160 £404.81
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
13495619 Memmert™
B29746
Model 30 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
13485619 Memmert™
B29751
Model 260 £278.56
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
13475619 Memmert™
B29753
Model 450 £473.92
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
11711484 Memmert™
B29749
Models 110 and 160 £272.90
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
11721484 Memmert™
B29747
Models 55 and 75 £231.69
Each View alternative products
Save Up to 
»
Estimated Shipment
23-07-2019
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Adjustable Subframe
  • Memmert Oven size 30 to 75: height 600mm
  • Size 110 to 450: height 500mm
Specifications

Specifications

Subframe