missing translation for 'onlineSavingsMsg'
Learn More

HUBER Control Cables

Cable Length: 3 m

Brand:  HUBER 6146

Code : 04

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 10073592

  • £168.30 / Each

Estimated Shipment: 21-02-2022
Add to basket

Description

Description

Cable Length: 3m
Specifications

Specifications

3 m
Tnt&Cc
Control Cable
Videos
SDS
Documents
Certificates
Special Offers

Special Offers