Learn More
Contec Wipe presaturated, low linting, center-pull canister, 50% polyester and 50% rayon, 70/30 IPA/DI 139.7mm x 203.2mm pack of 1,080 (6 x canister of 180 wipes) SATWipes™
Brand:  Contec SWNW0014
Wipe presaturated, low linting, center-pull canister, 50% polyester and 50% rayon, 70/30 IPA/DI 139.7mm x 203.2mm pack of 1,080 (6 x canister of 180 wipes) SATWipes™
Code : 04
Additional Details : Weight : 10.00000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Product Code. 15177915