missing translation for 'onlineSavingsMsg'
Learn More

Connexin 43 Mouse anti-Mouse, eFluor 660, Clone: Connexin43 (1A), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-4759-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15589946

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The Connexin43 monoclonal antibody recognizes mouse Connexin 43. Connexins are a family of integral membrane proteins that form gap junction channels, which function to facilitate direct cell-cell communication. Gap junctions are comprised of two hemichannels, or connexons, which are made up of six connexin proteins. Over 20 connexin proteins have been identified and each combine to form either homomeric or heteromeric channels with different functional properties. Connexin 43 is expressed in many tissues, including the central nervous system, heart, and bone. Connexin 43 is the most abundant connexin found in the myocardium and is also the primary connexin expressed in astrocytes. Mutations in connexin 43 are associated with ocluodentodigital dysplasia, a rare developmental disorder characterized by craniofacial and limb abnormalities. Connexin 43 may also be a promising therapeutic target in the treatment of malignant melanomas and mammary gland tumors.

The monoclonal antibody Connexin43 also recognizes human and rat connexin 43.
Specifications

Specifications

Connexin 43
Connexin43 (1A)
eFluor 660
Conjugated
IgG1, kappa
RUO
Primary
Human, Murine, Rat
Immunocytochemistry, Immunohistochemistry (Frozen), Microscopy
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Videos
Documents

For Research Use Only.