Learn More
Connexin 32 Monoclonal Antibody (Connexin32 (2A)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£104.00 - £343.00
Specifications
Antigen | Connexin 32 |
---|---|
Clone | Connexin32 (2A) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15537646
|
Affymetrix eBioscience
14-9759-82 |
100 μg |
£343.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15527646
|
Affymetrix eBioscience
14-9759-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
Connexin 32 | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
GJB1 | |
Primary | |
Antibody |
Connexin32 (2A) | |
Unconjugated | |
Mouse | |
Chicken, Human, Mouse, Rat | |
14618, 2705, 29584, 378797 | |
IgG1 κ | |
Affinity chromatography | |
GJB1 |
For Research Use Only.