missing translation for 'onlineSavingsMsg'
Learn More

Connexin 32, Biotin, clone: Connexin32 (2A), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9759-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15537376

  • £163.84 / 100µg


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The Connexin32 monoclonal antibody recognizes human, mouse, rat, and chicken Connexin 32. Connexins are a family of integral membrane proteins that form gap junction channels, which function to facilitate direct cell-cell communication. Gap junctions are comprised of two hemichannels, or connexons, which are made up of six connexin proteins. Over 20 connexin proteins have been identified and each combine to form either homomeric or heteromeric channels with different functional properties. Connexin 32 is expressed by myelin, neurons, oligodendrocytes, and hepatocytes, and plays a role in the development of ovarian follicles. Connexin 32 expression also plays a role in disease progression in chronic multiple sclerosis and experimental autoimmune encephalomyelitis. Mutations in the Connexin 32 gene, GJB1, result in a form of Charcot-Marie-Tooth disease which is characterized by peripheral nerve dysfunction.Applications Reported: This Connexin32 antibody has been reported for use in western blotting, immunocytochemistry, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This Connexin32 antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Connexin 32
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P08033, P28230, P08034
CMTX, CMTX1, CX32
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Western Blot
Connexin32 (2A)
Biotin
GJB1
Conjugated
Gjb1
IgG1, kappa
100μg
Store at 2-8°C. Do not freeze.
Primary
2705
Videos

For Research Use Only.