missing translation for 'onlineSavingsMsg'
Learn More

Connexin 32 Mouse, Alexa Fluor 488, Clone: Connexin32 (2A), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-9759-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15520287

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

The Connexin32 monoclonal antibody recognizes human, mouse, rat, and chicken Connexin 32. Connexins are a family of integral membrane proteins that form gap junction channels, which function to facilitate direct cell-cell communication. Gap junctions are comprised of two hemichannels, or connexons, which are made up of six connexin proteins. Over 20 connexin proteins have been identified and each combine to form either homomeric or heteromeric channels with different functional properties. Connexin 32 is expressed by myelin, neurons, oligodendrocytes, and hepatocytes, and plays a role in the development of ovarian follicles. Connexin 32 expression also plays a role in disease progression in chronic multiple sclerosis and experimental autoimmune encephalomyelitis. Mutations in the Connexin 32 gene, GJB1, result in a form of Charcot-Marie-Tooth disease which is characterized by peripheral nerve dysfunction.
Specifications

Specifications

Connexin 32
Connexin32 (2A)
Alexa Fluor 488
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Microscopy
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gap junction protein beta-1
IgG1, kappa
RUO
Primary
Rat
Videos

For Research Use Only.