Learn More
DWK Life Sciences DURAN™ Blank for separating funnel, conical for ISO 4800
For further processing as separating funnel with tap and standard ground joint.
Brand: DWK Life Sciences 210136302
Code : NEW
Additional Details : Weight : 1.35000kg
Description
DURAN™ separating funnels are used in chemistry to separate non-mixable liquids. DURAN™ separating funnels Raw bodies are designed for forming the appropriate standard ground joint sleeve according to DIN ISO 4800.
- Conical shape for ISO 4800
- Glass articles for further processing
Specifications
166 mm | |
Boro 3.3 | |
Blank for further processing as dropping funnel; Phase separation |
2000 mL | |
Yes | |
2/Pk. |