missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Conical Shape PTFE Key Separating Funnels

Brand:  Duran™ 1064805

Code : A0

Additional Details : Weight : 0.12300kg

 View more versions of this product

Product Code. 10580773

  • £67.00 / Each


Estimated Shipment
21-05-2018
Add to basket

Specifications

Specifications

9mm
100mL
Borosilicate Glass
Separating funnel
230mm
Videos