Learn More
DWK Life Sciences KECK™ Clip Assortment, for conical joints, from stainless steel (1.4310, blank)
The KECK™ clamp is used to secure a standard ground joint connection.
Brand: DWK Life Sciences 290300202
Code : NEW
Additional Details : Weight : 0.01100kg
Description
KECK™ clamps are ground joint clamps used to secure standard ground joint connections in glass apparatus and are characterized by their easy handling and excellent resilience. The clamps are available in different materials (metal or polyoxymethylene).
- Dimensionally stable with particularly good spring properties
- Easy, safe and quick to set up
Specifications
10 | |
Secure Glass-to-Glass Joints | |
NS 7 |
Stainless Steel (1.4310, Blank) | |
Secure glass-to-glass joints |