Learn More
Fistreem International™ Condenser
Replacement condenser for the Calypso or Cyclon Ultrapure Water Still. Fistreem International™ Condenser cools the steam produced in your system.
Brand: Fistreem International™ WSC705.CXJ.9
Code : 11
Additional Details : Weight : 1.13400kg
Description
- Borosilicate glass construction.
- Suitable for Calypso or Cyclon Ultrapure Water Still.
Specifications
1 kg | |
Calypso or Cyclon Ultrapure Water Still | |
Condenser | |
Borosilicate Glass |