missing translation for 'onlineSavingsMsg'
Learn More

Zeiss™ Condenser

Use this condenser for a variety of applications

Brand:  Zeiss™ 424228-9060-000

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 13488928

  • £59.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Condenser, achromatic-aplanatic 0.9 H with front lens which can be switched on the right and left
  • Condenser, achromatic-aplanatic 0.9 H D Ph DIC with front lens switchable on the left and right and 5-position modulator disk
  • Condenser 0.9/ 1.25 H, D, Ph1, Ph2, Ph3 for Axio Lab.A1 with low-power system from 2.5x
  • LD condenser 0.4 for slider; For H, Ph, PlasDIC with 10 x 46mm slider mount
Specifications

Specifications

Rotatable, with click stops 0°, 90°, 180°, 270°
LD condenser 0.4 for slider; For H, Ph, PlasDIC
Objectives 5x
0.4mm
1Each
Condenser
10 x 46mm slider mount
53mm
Videos