Learn More
3M™ Comply™ Indicator Tape
Enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. 3M™ Comply™ Indicator tape changes colour or turns darker when exposed to the sterilisation process.
Brand: 3M™ 7000138229
Code : NEW
Additional Details : Weight : 3.60000kg
Description
As an exposure monitor, 3M™ indicator tapes securely seal packs and enables CSSD and OR personnel to know at a glance that the packs have been exposed to the sterilisation process. The chemical indicator ink, printed in a diagonal striped pattern, changes colour or turns darker when exposed to the sterilisation process.- Lead-free steam indicator tape is designed for use on all wraps
- Reliable achesive adheres to packaging material
- Sustainable, solvent-free manufacturing proces
- Does not require hazardous waste disposal
- Redesigned packaging uses up to 30% less plastic
- Convenient, individually wrapped, easy-to-open rolls
- Lead free and no added latex
- 28 rolls/case
Specifications
Meets NF EN 867-1 Class A, ISO 11140-1 Class 1 | |
55 m x 18 mm | |
Autoclave | |
Indicator Tape | |
28 Pk. | |
Steam |
Beige | |
Sterilization verification | |
55 m | |
Lead-free, aggressive, pressure sensitive adhesive | |
Yes | |
18 mm |