missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Compact Jerrycan, Electroconductive

The compact jerrycan, electroconductive is especially well suited for use in explosion-proof areas. With a specific surface resistance of <105 ohms, the canister has a very high electrical conductivity.

£39.45 - £54.15

Specifications

Material HDPE, anti-static
For Use With (Application) For use in explosion-proof areas
Color Black
Closure Material HDPE
View More Specs

Products 4
Product Code Brand Capacity (Metric) Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Type Price Quantity & Availability  
16399531 View Documents Buerkle™
0435-1055
5 L Jerry Can £39.45
Each View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
16309541 View Documents Buerkle™
0435-1105
10 L Jerry Can £52.45
Each View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
16319541 View Documents Buerkle™
0435-2055
5 L Jerry Can £41.20
Each View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
16329541 View Documents Buerkle™
0435-2105
10 L Jerry Can £54.15
Each View alternative products
Save Up to 
»
Estimated Shipment
12-02-2020
Add to basket
 
Description

Description

  • HDPE, black, anti-static
  • For use in explosion-proof areas
  • For dispensing and storing combustible and highly flammable liquids
  • With or without threaded connector 3/4 in.
  • Due to its narrow width, the canister requires very little room
Specifications

Specifications

HDPE, anti-static
For use in explosion-proof areas
Black
HDPE