missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Common gamma Chain/IL-2 R gamma Antibody (31134) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2841JF525
This item is not returnable.
View return policy
Description
Common gamma Chain/IL-2 R gamma Monoclonal antibody specifically detects Common gamma Chain/IL-2 R gamma in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Common gamma Chain/IL-2 R gamma | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD132, CD132 antigen, CIDX, combined immunodeficiency, X-linked, common cytokine receptor gamma chain, common gamma-chain, cytokine receptor common subunit gamma, gamma(c), IL-2 receptor subunit gamma, IL-2R subunit gamma, IL-2RG, IMD4, interleukin 2 receptor, gamma, Interleukin-2 receptor subunit gamma, p64, SCIDX, SCIDX1, severe combined immunodeficiency | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Common gamma Chain/IL-2 R gamma , Leu23-Asn254, Accession # P31785 | |
| 0.5 mL | |
| Cancer | |
| 3561 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 31134 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction