Learn More
Fisherbrand™ Combined pH Electrode with BNC Connector
Combined pH electrode with BNC connector is ideal for use in water analysis.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.80000kg
Description
- Used in water analysis
- PT1000 probe
- BNC connector
- Combined pH electrode type
Specifications
BNC | |
Combined pH Electrode |