Learn More
WTW™ Combination pH electrode SensoLyt SEA
Brand: WTW™ 103710
Code : 03
Additional Details : Weight : 0.10000kg
Specifications
TA 197-pH, Polymer Electrolyte, Pin Hole Junction | |
For installation in depth armature series and for heavily polluted waters | |
Pin Hole |
Combination pH Electrode SensoLyt SEA | |
Built-metric temperature sensor and gel electrolyte | |
Polymer |