Learn More
Fisherbrand™ Comb Accessory
Three different Vari-gel tanks, 150mm, 100mm and 70mm are available accommodating up to four different gel tray sizes
Brand:  Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Product Code. 12329778
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Three different Vari-gel tanks, 150mm, 100mm and 70mm are available accommodating up to four different gel tray sizes
  • UV transparent gel trays allow for placement directly onto a UV transilluminator for gel viewing once stained
  • Asymmetric electrode offset - prevents accidental polarity reversal ensuring no possibility of sample loss into the buffer
  • aluminum casting gates - ensure easy casting of the gel inside or outside of the tank. They slide into pre-formed grooves at the end of the gel tray providing a secure seal against gel leakage
  • Different gel sizes can be interchanged in one tank, combining superb resolution with ease of use.
Specifications
Specifications
Comb
8
1 Each
1 mm