missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 0.75mm Thick
For use in preparatory electrophoresis and high-throughput screening.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.01000kg
Description
- For tightly resolved bands
- Recommended for high resolution gels and publication quality data
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
4 Prep, 2 Marker | |
91 μL |
Comb | |
0.75 mm |