Learn More
Brady™ Color Polyester Laboratory Vial and Tube Labels
Easily differentiate lab samples with these B-494 color polyester labels, which are available in six color options. Pair with a versatile Brady™ portable label printer and print almost anywhere.
Brand:  Brady™ 170917
Additional Details : Weight : 0.16100kg
 View more versions of this product
Product Code. 17831779
  • £60.78 / Pack of 240
Estimated Shipment: 23-09-2024
to see stock.
Description
Description
  • Ideal for easily differentiating vials, PCR and microcentrifuge tubes, and slides
  • Designed to withstand abrasions and exposure to chemicals and extreme temperatures, including immersion in liquid nitrogen and hot water baths
  • Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces such as glass and polypropylene
  • Available in six color options with a color bar across the top of the label; pre-sized for multiple applications, some options with matching color vial top label
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Gloss
Polyester
Labels
Cold Temperature Labeling, Laboratory Labeling
240 Labels/Cartridge
0.003 in.
0.5 in.
2.54 cm
0.375 in.
RoHS Compliant to 2011/65/EU (Pass 2015/863)
Pink, White
Die-cut
Thermal Transfer
Non-sterile
BMP51, M511
-320.8°F to +158°F
0.5 x 1 in.
1 in.
0.5 in.