missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Coded Buffer Solution pH 7, SI Analytics™
Click to view available options
Quantity:
250 mL
Packaging:
Plastic Bottle
Description
- The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
- Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
- Color coded
Specifications
Boiling Point | 100°C |
pH | 7 |
Physical Form | Liquid |
Quantity | 250 mL |
Packaging | Plastic Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'