Learn More
Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 41-9771-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.
Collagen X Monoclonal antibody specifically detects Collagen X in Canine, Guinea Pig, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Collagen X | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q03692, Q05306 | |
COL10A1 | |
Affinity chromatography | |
RUO | |
100856711, 106026924, 12813, 1300, 25681, 448809 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
X53 | |
eFluor 570 | |
COL10A1 | |
Col10; COL10A1; Col10a-1; Collagen alpha-1(X) chain; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1 | |
Mouse | |
25 μg | |
Primary | |
Canine, Guinea Pig, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG1 κ |
For Research Use Only.