missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen X Monoclonal Antibody (X53), eFluor™ 570, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£193.00 - £447.00
Specifications
Antigen | Collagen X |
---|---|
Clone | X53 |
Concentration | 0.2 mg/mL |
Applications | Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15588966
|
Affymetrix eBioscience
41-9771-80 |
25 μg |
£193.00
25µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15598966
|
Affymetrix eBioscience
41-9771-82 |
100 μg |
£447.00
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.
Collagen X Monoclonal antibody specifically detects Collagen X in Canine, Guinea Pig, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
Collagen X | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Col10; COL10A1; Col10a-1; Collagen alpha-1(X) chain; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1 | |
COL10A1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
COL10A1 |
X53 | |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
eFluor 570 | |
Mouse | |
Canine, Guinea Pig, Human, Mouse, Porcine, Rat | |
Q03692, Q05306 | |
100856711, 106026924, 12813, 1300, 25681, 448809 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.