missing translation for 'onlineSavingsMsg'
Learn More

Collagen X Monoclonal Antibody (X53), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9771-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15577666

  • £405.00 / 100µg
Estimated Shipment: 23-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody X53 recognizes human, mouse, porcine, guinea pig, and rat Collagen Type X. X53 does not cross-react with collagen types I, II, III, IV, or VI. Collagen Type X is a homotrimeric short chain collagen. Collagen Type X has limited expression in normal adult tissues, primarily being expressed during bone growth and development. Expression is localized to the peri- and extracellular matrix of hypertrophic chondrocytes during endochondral ossification and also plays a role in cartilage mineralization. Collagen Type X is elevated in many tumor types with expression being localized in the vasculature. Mutations to the gene COL10A1, which encodes human Collagen Type X, is associated with Schmid type metaphyseal chondrodysplasia. Applications Reported: This X53 antibody has been reported for use in microscopy, immunohistochemical staining, ELISA, and immunocytochemistry. Applications Tested: This X53 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Type X collagen is a product of hypertrophic chondrocytes and has been localized to presumptive mineralization zones of hyaline cartilage.
Specifications

Specifications

Collagen X
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q03692, Q05306
COL10A1
Affinity chromatography
RUO
100856711, 106026924, 12813, 1300, 25681, 448809
4° C
Liquid
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin)
X53
Unconjugated
COL10A1
Col10; COL10A1; Col10a-1; Collagen alpha-1(X) chain; collagen type X alpha 1 chain; collagen X, alpha-1 polypeptide; collagen, type X, alpha 1; collagen, type X, alpha 1(Schmid metaphyseal chondrodysplasia); pro alpha 1; procollagen, type X, alpha 1; Schmid metaphyseal chondrodysplasia; type X collagen; type X collagen, alpha 1
Mouse
100 μg
Primary
Canine, Guinea Pig, Human, Mouse, Porcine, Rat
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.