missing translation for 'onlineSavingsMsg'
Learn More

Collagen IV, eFluor 660, clone: 1042, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9871-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15530167

  • £162.23 / 100µg


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Description

Description

Description: This monoclonal antibody 1042 recognizes human collagen Type IV. Collagens are the main components of the extracellular matrix and provide the tensile strength to tissues. There are four main types of collagen (I, II, III and IV) which contribute to the connective tissue. Expression of type IV is restricted to the basal lamina. It contains two alpha-1 and one alpha-2-like chains. Collagen Type IV is associated with disorders such as Alport syndrome and Goodpasture's syndrome.Applications Reported: This 1042 antibody has been reported for use in immunohistochemical staining, immunohistochemical staining of frozen tissue sections, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This 1042 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using low pH antigen retrieval and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Collagen Type IV
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P02462
COL4A1
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1282
Immunohistochemistry (Frozen)
1042
eFluor 660
COL4A1
Conjugated
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Videos

For Research Use Only.