missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Collagen, Bovine Achilles tendon

For research use

Brand:  Alfa Aesar™ J60218.03

UNSPSC : 12352200

Code : NEW

Additional Details : CAS Number : 9007-34-5 Weight : 0.00100kg

 View more versions of this product

Product Code. 15493019

  • £32.65 / 1g


Estimated Shipment date
from Supplier 09-06-2017
Add to basket

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

An attachment factor for primary cultures of epithelioid cells
Lyophilized powder
Bovine achilles tendon
RUO
No
Lyophilized Powder
1g
2° to 8°C
Collagen, Bovine achilles tendon
Documentation

Technical Documents