missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Cole-Parmer™ Masterflex™ L/S™ Platinum-Cured Silicone High-Performance Precision Tubing

Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Platinum-Cured Silicone High-Performance Precision Tubing provides accurate, repeatable flow. Ideal for Pharmaceutical, biotech, peristaltic pumping and food applications.

Brand:  Cole-Parmer™ 96410-24

Code : XX

Additional Details : Weight : 0.85070kg

 View more versions of this product

Product Code. 11875181

  • £171.47 / 7.60m


In Stock 1
Add to basket

Description

Description

  • Manufactured to extremely close tolerances that match L/S™ pump heads, ensuring accurate, repeatable flow and long tubing life
  • Factory-tested and optically inspected to provide the best performance from your peristaltic pump
  • Features a thicker wall compared to our Precision pump tubing, making it the best choice for applications involving pressure, suction lift and viscous fluids
  • Fewer potential leachables
  • Excellent biocompatibility
  • Odorless and nontoxic, with no taste imparted to transported fluids
  • Resistant to fungus growth, weather, ozone, corona and radiation
  • Smoother surface than peroxide-cured silicone for reduced protein binding
  • Excellent heat resistance, low compression set, odorless, nontoxic
  • Translucent
Specifications

Specifications

Yes
2.8 to 1700mL/min.
Pump Tubing
For pharmaceutical, biotech, peristaltic pumping, and food applications
8.8m
230°C
–50°C
Translucent
Platinum-cured Silicone
760cm
Silicone (platinum-cured)
1/4 in.
40psi
24
Videos
Documentation